SY3H5815F5C4 Synthonix Corporation

(1-(tert-Butoxycarbonyl)-4-fluoro-1H-indol-2-yl)boronic acid

  • Cas Number : 1000068-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3592.html

Safety Information

MSDS https://synthonix.com/3592.html
Availability details for
Quantity
  • Availability
  • Add to Cart