SY3H5815F5CF Synthonix Corporation

7-Benzyl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione

  • Cas Number : 62459-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4273.html

Safety Information

MSDS https://synthonix.com/4273.html
Availability details for
Quantity
  • Availability
  • Add to Cart