SY3H5815F5DE Synthonix Corporation

7-Benzyl-4-chloro-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine

  • Cas Number : 192869-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4589.html

Safety Information

MSDS https://synthonix.com/4589.html
Availability details for
Quantity
  • Availability
  • Add to Cart