SY3H5815F5F0 Synthonix Corporation

tert-Butyl 1,6-diazaspiro[3.5]nonane-1-carboxylate

  • Cas Number : 1251005-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8311.html

Safety Information

MSDS https://synthonix.com/8311.html
Availability details for
Quantity
  • Availability
  • Add to Cart