SY3H5815F5F4 Synthonix Corporation

[2,2'-Bipyridine]-4,4'-diamine

  • Cas Number : 18511-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/8876.html

Safety Information

MSDS https://synthonix.com/8876.html
Availability details for
Quantity
  • Availability
  • Add to Cart