SY3H5815F643 Synthonix Corporation

1-Chloro-5-isopropoxy-2-methyl-4-nitrobenzene

  • Cas Number : 1032903-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16150.html

Safety Information

MSDS https://synthonix.com/16150.html
Availability details for
Quantity
  • Availability
  • Add to Cart