SY3H5815F645 Synthonix Corporation

4-Chloro-5-iodo-2-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • Cas Number : 1060815-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16345.html

Safety Information

MSDS https://synthonix.com/16345.html
Availability details for
Quantity
  • Availability
  • Add to Cart