SY3H5815F64A Synthonix Corporation

2-(Chloromethyl)-4-methylquinazoline

  • MDL number MFCD09807547
  • Cas Number : 109113-72-6
  • Molecular Weight : 192.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow red
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/16498.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16498.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart