SY3H5815F64C Synthonix Corporation

5-Chloropyrazolo[1,5-a]pyridine

  • Cas Number : 1101120-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16568.html

Safety Information

MSDS https://synthonix.com/16568.html
Availability details for
Quantity
  • Availability
  • Add to Cart