SY3H5815F64D Synthonix Corporation

6-Chloro-5-(trifluoromethyl)nicotinic acid

  • Cas Number : 1110782-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16608.html

Safety Information

MSDS https://synthonix.com/16608.html
Availability details for
Quantity
  • Availability
  • Add to Cart