SY3H5815F64E Synthonix Corporation

6-Chloro-2,5-dimethylpyridazin-3(2H)-one

  • Cas Number : 1114563-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16622.html

Safety Information

MSDS https://synthonix.com/16622.html
Availability details for
Quantity
  • Availability
  • Add to Cart