SY3H5815F653 Synthonix Corporation

2-Chloro-3-nitro-6-(trifluoromethyl)pyridine

  • Cas Number : 117519-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16978.html

Safety Information

MSDS https://synthonix.com/16978.html
Availability details for
Quantity
  • Availability
  • Add to Cart