SY3H5815F654 Synthonix Corporation

1-Cyclohexyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • Cas Number : 1175275-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16982.html

Safety Information

MSDS https://synthonix.com/16982.html
Availability details for
Quantity
  • Availability
  • Add to Cart