SY3H5815F660 Synthonix Corporation

3-Chloro-6-methoxypyrazin-2-amine

  • Cas Number : 13484-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18840.html

Safety Information

MSDS https://synthonix.com/18840.html
Availability details for
Quantity
  • Availability
  • Add to Cart