SY3H5815F668 Synthonix Corporation

1-Cyclopropyl-2-(2-fluorophenyl)ethanone

  • Cas Number : 150322-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20123.html

Safety Information

MSDS https://synthonix.com/20123.html
Availability details for
Quantity
  • Availability
  • Add to Cart