SY3H5815F669 Synthonix Corporation

2-Chloro-4-nitropyridin-3-ol

  • Cas Number : 15128-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20163.html

Safety Information

MSDS https://synthonix.com/20163.html
Availability details for
Quantity
  • Availability
  • Add to Cart