SY3H5815F675 Synthonix Corporation

7-Chloro-N-(3-chloro-4-fluorophenyl)-6-nitroquinazolin-4-amine

  • Cas Number : 179552-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21921.html

Safety Information

MSDS https://synthonix.com/21921.html
Availability details for
Quantity
  • Availability
  • Add to Cart