SY3H5815F681 Synthonix Corporation

4-Chloro-6-nitroquinazoline

  • Cas Number : 19815-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22135.html

Safety Information

MSDS https://synthonix.com/22135.html
Availability details for
Quantity
  • Availability
  • Add to Cart