SY3H5815F68F Synthonix Corporation

4-Chloro-N-methylpyrimidin-2-amine

  • Cas Number : 22404-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22466.html

Safety Information

MSDS https://synthonix.com/22466.html
Availability details for
Quantity
  • Availability
  • Add to Cart