SY3H5815F69D Synthonix Corporation

(S)-N4-(3-Chloro-4-fluorophenyl)-7-((tetrahydrofuran-3-yl)oxy)quinazoline-4,6-diamine

  • Cas Number : 314771-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22995.html

Safety Information

MSDS https://synthonix.com/22995.html
Availability details for
Quantity
  • Availability
  • Add to Cart