SY3H5815F6A0 Synthonix Corporation

6-Chloro-2-methoxypyrimidin-4-amine

  • Cas Number : 3286-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23069.html

Safety Information

MSDS https://synthonix.com/23069.html
Availability details for
Quantity
  • Availability
  • Add to Cart