SY3H5815F6A2 Synthonix Corporation

1-Chloro-4-methoxyisoquinoline

  • Cas Number : 3336-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23098.html

Safety Information

MSDS https://synthonix.com/23098.html
Availability details for
Quantity
  • Availability
  • Add to Cart