SY3H5815F6A4 Synthonix Corporation

1-Chloro-2,5-dimethyl-4-nitrobenzene

  • MDL number MFCD00087057
  • Cas Number : 34633-69-7
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/23187.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/23187.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart