SY3H5815F6AC Synthonix Corporation

2-Chloro-8-methylquinoline

  • Cas Number : 4225-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23632.html

Safety Information

MSDS https://synthonix.com/23632.html
Availability details for
Quantity
  • Availability
  • Add to Cart