SY3H5815F6C4 Synthonix Corporation

2-Chloro-5-nitroimidazole

  • Cas Number : 57531-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24345.html

Safety Information

MSDS https://synthonix.com/24345.html
Availability details for
Quantity
  • Availability
  • Add to Cart