SY3H5815F6CA Synthonix Corporation

4-Chloro-3-methylquinoline

  • Cas Number : 63136-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24644.html

Safety Information

MSDS https://synthonix.com/24644.html
Availability details for
Quantity
  • Availability
  • Add to Cart