SY3H5815F6D5 Synthonix Corporation

5-Chloroisoindoline-1,3-dione

  • Cas Number : 7147-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25030.html

Safety Information

MSDS https://synthonix.com/25030.html
Availability details for
Quantity
  • Availability
  • Add to Cart