SY3H5815F6D6 Synthonix Corporation

2-Chloro-5-methoxypyridin-3-amine

  • Cas Number : 720666-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25045.html

Safety Information

MSDS https://synthonix.com/25045.html
Availability details for
Quantity
  • Availability
  • Add to Cart