SY3H5815F6D8 Synthonix Corporation

2-(Chloromethyl)-4,6-dimethylpyrimidine

  • Cas Number : 74502-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25169.html

Safety Information

MSDS https://synthonix.com/25169.html
Availability details for
Quantity
  • Availability
  • Add to Cart