SY3H5815F6DB Synthonix Corporation

3-Chlorobenzo[d]isothiazole

  • Cas Number : 7716-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25325.html

Safety Information

MSDS https://synthonix.com/25325.html
Availability details for
Quantity
  • Availability
  • Add to Cart