SY3H5815F6E1 Synthonix Corporation

6-(Chloromethyl)nicotinonitrile

  • Cas Number : 83640-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25620.html

Safety Information

MSDS https://synthonix.com/25620.html
Availability details for
Quantity
  • Availability
  • Add to Cart