SY3H5815F6E4 Synthonix Corporation

2-Chloro-6-(trifluoromethyl)pyrimidin-4-amine

  • Cas Number : 85730-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25741.html

Safety Information

MSDS https://synthonix.com/25741.html
Availability details for
Quantity
  • Availability
  • Add to Cart