SY3H5815F6E6 Synthonix Corporation

4-Chloro-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 869335-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25914.html

Safety Information

MSDS https://synthonix.com/25914.html
Availability details for
Quantity
  • Availability
  • Add to Cart