SY3H5815F6E8 Synthonix Corporation

(R)-2-Chloro-7-ethyl-8-isopropyl-5-methyl-7,8-dihydropteridin-6(5H)-one

  • Cas Number : 877676-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26021.html

Safety Information

MSDS https://synthonix.com/26021.html
Availability details for
Quantity
  • Availability
  • Add to Cart