SY3H5815F6F4 Synthonix Corporation

2-Chlorothiazolo[5,4-b]pyridine

  • Cas Number : 91524-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26456.html

Safety Information

MSDS https://synthonix.com/26456.html
Availability details for
Quantity
  • Availability
  • Add to Cart