SY3H5815F702 Synthonix Corporation

1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride

  • Cas Number : 2031-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3154.html

Safety Information

MSDS https://synthonix.com/3154.html
Availability details for
Quantity
  • Availability
  • Add to Cart