SY3H5815F70E Synthonix Corporation

6-Chloropyrazine-2-carbonitrile

  • Cas Number : 6863-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/4892.html

Safety Information

MSDS https://synthonix.com/4892.html
Availability details for
Quantity
  • Availability
  • Add to Cart