SY3H5815F732 Synthonix Corporation

Dimethyl pyridine-3,5-dicarboxylate

  • Cas Number : 4591-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0837.html

Safety Information

MSDS https://synthonix.com/0837.html
Availability details for
Quantity
  • Availability
  • Add to Cart