SY3H5815F740 Synthonix Corporation

2,7-Diazaspiro[4.5]decan-1-one hydrochloride

  • Cas Number : 1187173-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17070.html

Safety Information

MSDS https://synthonix.com/17070.html
Availability details for
Quantity
  • Availability
  • Add to Cart