SY3H5815F74F Synthonix Corporation

3,5-Dichloro-4-formylbenzoic acid

  • Cas Number : 153203-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20301.html

Safety Information

MSDS https://synthonix.com/20301.html
Availability details for
Quantity
  • Availability
  • Add to Cart