SY3H5815F752 Synthonix Corporation

2,3-Dibromo-5-nitropyridine

  • Cas Number : 15862-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20503.html

Safety Information

MSDS https://synthonix.com/20503.html
Availability details for
Quantity
  • Availability
  • Add to Cart