SY3H5815F754 Synthonix Corporation

(2,6-Dichloro-3-nitrophenyl)methanol

  • Cas Number : 160647-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/20551.html

Safety Information

MSDS https://synthonix.com/20551.html
Availability details for
Quantity
  • Availability
  • Add to Cart