SY3H5815F759 Synthonix Corporation

2,6-Dibromo-4-nitropyridine

  • Cas Number : 175422-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21865.html

Safety Information

MSDS https://synthonix.com/21865.html
Availability details for
Quantity
  • Availability
  • Add to Cart