SY3H5815F75B Synthonix Corporation

1-(3,4-Dimethylphenyl)-3-methyl-1H-pyrazol-5(4H)-one

  • Cas Number : 18048-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21933.html

Safety Information

MSDS https://synthonix.com/21933.html
Availability details for
Quantity
  • Availability
  • Add to Cart