SY3H5815F75F Synthonix Corporation

3,5-Difluoro-4-(trifluoromethoxy)phenol

  • Cas Number : 195206-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22106.html

Safety Information

MSDS https://synthonix.com/22106.html
Availability details for
Quantity
  • Availability
  • Add to Cart