SY3H5815F763 Synthonix Corporation

3,5-Dibromo-1-methyl-1H-1,2,4-triazole

  • Cas Number : 23579-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22545.html

Safety Information

MSDS https://synthonix.com/22545.html
Availability details for
Quantity
  • Availability
  • Add to Cart