SY3H5815F76C Synthonix Corporation

3,5-Dichloro-6-ethylpyrazine-2-carboxamide

  • Cas Number : 313340-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22981.html

Safety Information

MSDS https://synthonix.com/22981.html
Availability details for
Quantity
  • Availability
  • Add to Cart