SY3H5815F771 Synthonix Corporation

3,4-Diaminopyridin-2-ol

  • Cas Number : 33631-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23116.html

Safety Information

MSDS https://synthonix.com/23116.html
Availability details for
Quantity
  • Availability
  • Add to Cart