SY3H5815F772 Synthonix Corporation

3,6-Dichloro-4,5-dimethylpyridazine

  • Cas Number : 34584-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23184.html

Safety Information

MSDS https://synthonix.com/23184.html
Availability details for
Quantity
  • Availability
  • Add to Cart