SY3H5815F77D Synthonix Corporation

4,6-Dibromopyridin-2-amine

  • Cas Number : 408352-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23567.html

Safety Information

MSDS https://synthonix.com/23567.html
Availability details for
Quantity
  • Availability
  • Add to Cart